Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
G418 disulphate
G418 disulphate, powder
G418 disulphate, powder
G418 disulphate, powder
Supplier:
Corning
Certificates
Expand image
G418 disulphate, powder
Supplier:
Corning
Certificates
About this item
CAS Number:
108321-42-2
:
Formula:
C₂₀H₄₄N₄O₁₈S₂
MDL Number:
MFCD00058314
:
692.72 g/mol
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Corning Cell culture media and reagents - Product selection guide
Documentation