VWR® pHenomenal® pH Electrodes
Supplier: VWR Collection
Certificates
About this item
pH electrodes with BNC connector and cable.
These electrodes are checked at five points during the manufacturing process and then lot stamped to ensure quality. Every electrode is supplied with a soaker storage bottle and cap to ensure the pH-sensitive glass and reference junction remain in top condition and are instantly ready for use.
* Integrated temperature probe.
Delivery: Includes pH-electrode only. 620-1659 includes pHenomenal® TEMP21 T-sensor (NTC 30 kOhm) only.
Related Information
CE Certificate_VWR Electrode.pdf CE_VWR_Phenomenal_pH_electrode_Conductivity_oxygen_meter.pdf EUDoC_portal_info
Manual VWR® Phenomenal pH Sensors Manual_VWR_IS_2000_L_Bedienungsanleitung_DE_270213 Manual_VWR_IS_2000_L_e_operativo_IT_270213 Manual_VWR_IS_2000_L_Instrucciones_de_operacion_ES_270213 Manual_VWR_IS_2000_L_Instrucoes_de_Utilizacao_PT_270213
Manual
More
VWR Electrochemistry VWR pH Buffer Solutions VWR pH Electrodes
Product Literature
VWRI662-1161_TDS_EN