Order Entry
Finland
ContactUsLinkComponent
Formaldehyde 4% stabilised, buffered (pH 7.0 ± 0.2)
Health
Irritant
Formaldehyde 4% stabilised, buffered (pH 7.0 ± 0.2)
Catalog # VWRC6370.9200
Supplier:  VWR Chemicals
CAS Number:  
Health
Irritant
Formaldehyde 4% stabilised, buffered (pH 7.0 ± 0.2)
Catalog # VWRC6370.9200
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    200 L
  • Seq:
    5549

Specifications

About this item

 :
 :30.03 g/mol
 :Stabilised with methanol 1%
 :GHS08
 :GHS07
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274