Spatula, Hayman type
NICK9570
- Length:165 mm
- Blade width:3 / 3,5 mm
- Pk:1
Stainless steel 18/8, non magnetic, bright finish.
- Corrosion resistant and durable
- Suitable for intermittent use up to 870 °C
This spatula has a 3 mm wide curved half moon end at the tip. This end is sharpened at the tip for cutting, separating or handling mico samples. The other end features a flat blade for handling small samples (3,5 mm wide).