Striped safety cleanroom tapes, SST™
Supplier: MICRONOVA
Certificates
About this item
These pressure sensitive laminated tapes are made of vinyl with rubber adhesive and are ideal for delineating and colour coding specific equipment and areas.
- Tape is scuff resistant and holds up well to solvents, humidity and oil
- Wound on a plastic core and cleanroom processed
Length: 16,4 m
Different colours available.
Colour: Black and yellow striped