- Antibody type:Primary
- Antigen name:B melanoma antigen 2
- Antigen symbol:BAGE2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:BAGE2_HUMAN.|member 2|B melanoma antigen family|CT2.2|Cancer/testis antigen 2.2|B melanoma antigen 2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Members of the BAGE gene family encode antigens that are recognized by cytotoxic T lymphocytes and are also known as CT (cancer/testis) antigens. Generated by juxtacentromeric shuffling of the MLL3 gene, the ancestral BAGE gene was expanded by acrocentric exchanges and/or juxtacentromeric movements.Generally, BAGE proteins are silent in all normal tissues with the exception of testis. BAGE2 and BAGE 3 (B melanoma antigen 2 and 3, respectively), also known as Cancer/testis antigen 2.2 and 2.3 (respectively), are 109 amino acid secreted proteins that are expressed in 22% of melanomas, lung and bladder carcinomas, and are also expressed in normal testis tissue. Like the genes encoding MAGE proteins, BAGE genes are most likely silenced by DNA methylation and/or chromatin compaction in normal tissues other than testis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BAGE2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human