Anti-TSPEAR Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-9577R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Thrombospondin-type laminin G Domain and EAR repeats
- Antigen symbol:TSPEAR
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:TSP-EAR|DFNB98|C21orf29
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
TSPEAR, also known as C21orf9, is a 669 amino acid secreted protein. Expressed as two isoforms produced by alternative splicing, TSPEAR contains one Thrombospondin N-terminal domain and seven EAR (epilepsy-associated repeat) domains. EAR domains are found in several proteins, including TSPEAR, encoded by genes that map within chromosome regions associated with seizure disorders. It is thought that the EAR domain plays a role in the pathogenesis of epilepsy by either binding to an unknown epileptic ligand or interfering with axon synaptogenesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TSPEAR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat