Specifications
- Antibody type:Primary
- Antigen name:Scavenger Receptor Class B, member 2
- Antigen symbol:SCARB2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:950
- Antigen synonyms:LIMP-2|EPM4|CD36L2|LIMP2|AMRF|85 kDa lysosomal membrane sialoglycoprotein|LIMPII|SR-BII|LGP85|Lysosome membrane protein 2|HLGP85
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:222-242/478
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. SR-BI and SR-BII (previously known as SR-BI.2) are the alternatively spliced products of a single gene. SR-BII and SR-BI are identical except for the encoded c-terminal cytoplasmic domain. Both SR-BI and SR-BII bind HDL and mediates selective uptake of HDL cholesteryl ester, but with SR-BII having an approximately 4-fold lower efficiency than SR-BI. SR-BI and SR-BII are expressed primarily in liver and non-placental steroidgenic tissues. Although the role of these scavenger receptors is not completely clear, SR-BII mRNA results from the alternative splicing of SR-BI precursor transcripts with both isoforms mediating selective transfer of lipid between HDL and cells. Therefore, the relative expression and functional activities of these two isoforms create a potential means of regulating selective lipid transfer between HDL and cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SCARB2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat