Order Entry
Finland
ContactUsLinkComponent
Anti-SCAF1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-SCAF1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-6735R-A647
Supplier:  Bioss
Anti-SCAF1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-6735R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SR-Related CTD Associated Factor 1
  • Antigen symbol:
    SCAF1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    58506
  • Antigen synonyms:
    SFR19_HUMAN.|SR-related and CTD-associated factor 1|Splicing factor|Serine arginine-rich pre-mRNA splicing factor SR-A1|SR-related-CTD-associated factor|arginine/serine-rich 19|SR-A1|SCAF
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

SR-A1 (serine arginine-rich pre-mRNA splicing factor) is a member of the SR protein family, which interacts with the C-terminal domain of the large subunit of RNA polymerase II and involved in pre-mRNA splicing. The SR-A1 gene is located on chromosome 19q13.3-q13.4. Overexpression of SR-A1 is found in aggressive ovarian cancers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCAF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat