Anti-TR1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-6117R-A555
: Bioss
- Antibody type:Primary
- Antigen name:Tomoregulin 1
- Antigen symbol:TR1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:8577
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:75-105/380
- Purification:Purified by Protein A
- Pk:100 µl
May inhibit NODAL and BMP signaling during neural patterning (By similarity). May be a tumor suppressor in brain cancers.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat