Anti-HYAL2 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-5888R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Hyaluronidase 2
- Antigen symbol:HYAL2
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:8692
- Antigen synonyms:Lung carcinoma protein 2|Hyal-2|Hyaluronidase-2|LUCA2|HYAL2|LuCa-2|Hyaluronoglucosaminidase-2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:150-200/473
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Hydrolyzes high molecular weight hyaluronic acid to produce an intermediate-sized product which is further hydrolyzed by sperm hyaluronidase to give small oligosaccharides. Displays very low levels of activity. Associates with and negatively regulates MST1R.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HYAL2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat