Order Entry
Finland
ContactUsLinkComponent
Anti-NT5E Rabbit Polyclonal Antibody (Cy5.5®)
Anti-NT5E Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-4834R-CY5.5
Supplier:  Bioss
Anti-NT5E Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-4834R-CY5.5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5'-Nucleotidase, Ecto
  • Antigen symbol:
    NT5E
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Chicken,
    Human,
    Dog,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    4907
  • Antigen synonyms:
    eN|E5NT|NTE|NT5|CALJA|eNT|NT|CD73
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    208-222/574
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Hydrolyzes extracellular nucleotides into membrane permeable nucleosides. Exhibits AMP-, NAD-, and NMN-nucleosidase activities.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: NT5E
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Dog, Chicken