Specifications
- Antibody type:Primary
- Antigen name:DNA replication licensing factor MCM2
- Antigen symbol:MCM2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:4171
- Antigen synonyms:MITOTIN|minichromosome maintenance protein 2 homolog|cyclin-like 1|D3S3194|cell devision cycle-like 1|CCNL1|MCM2 minichromosome maintenance deficient 2|mitotin|minichromosome maintenance deficient 2 (mitotin)|DNA replication licensing factor MCM2|nuclear protein BM28|BM28|cdc19|CDCL1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:635-675/904
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Acts as component of the MCM2-7 complex (MCM complex) which is the putative replicative helicase essential for 'once per cell cycle' DNA replication initiation and elongation in eukaryotic cells. The active ATPase sites in the MCM2-7 ring are formed through the interaction surfaces of two neighboring subunits such that a critical structure of a conserved arginine finger motif is provided in trans relative to the ATP-binding site of the Walker A box of the adjacent subunit. The six ATPase active sites, however, are likely to contribute differentially to the complex helicase activity. Required for the entry in S phase and for cell division.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MCM2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat