Anti-HMP Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-1824R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Inner membrane Protein mitochondrial
- Antigen symbol:HMP
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10989
- Antigen synonyms:HMP|PIG4|P87|P89|P87/89|MINOS2|PIG52|Mic60
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:250-300/758
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintenance of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Plays an important role in the maintenance of the MICOS complex stability and the mitochondrial cristae morphology (PubMed:22114354, PubMed:25781180).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Mitofilin/IMMT
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat