Specifications
- Antibody type:Primary
- Antigen name:Exostosin-Like Glycosyltransferase 3
- Antigen symbol:EXTL3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2137
- Antigen synonyms:REGR|EXTR1|RPR|BOTV|EXTL1L
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
EXTL3 is a member of the EXT (hereditary multiple exostosin) gene family of tumor suppressors encoding glycosyltransferases involved in heparan sulfate (HS) biosynthesis. Within this family, the C-terminus is conserved between all members from C. elegans to vertebrates. EXTL3 is a ubiquitously expressed, developmentally regulated, single-pass type II membrane protein that localizes to the endoplasmic reticulum membrane. EXTL3 adds N-acetylglucosamine (GlcNAc) to the polysaccharide-protein linkage region and to the growing HS chain suggesting that it plays a role in both the initiation and elongation of HS chains. In addition, EXTL3 may act as a Reg receptor, binding Reg via its N-terminus.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EXTL3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat