Specifications
- Antibody type:Primary
- Antigen name:Attractin-Like 1
- Antigen symbol:ATRNL1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:bA338L11.1|ALP|bA454H24.1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
ATRNL1 is a 1,379 amino acid single-pass type I membrane protein that may play a role in melanocortin signaling pathways that regulate energy homeostasis. The ATRNL1 protein contains a C-type lectin domain, a CUB domain, two EGF-like domains, six Kelch repeats, two laminin EGF-like domains and five PSI domains. ATRNL1 interacts with MC4-R in several regions known to be important in the regulation of energy homeostasis by melanocortins, such as the paraventricular nucleus of hypothalamus and the dorsal motor nucleus of the vagus. The ATRNL1 gene is conserved in dog, cow, mouse, rat, chicken, zebrafish and C. elegans, exists as two alternatively spliced isoforms and maps to human chromosome 10q25.3. Strong evidence of linkage to late-onset Alzheimer disease (LOAD) is linked to chromosome 10, which implicates a wide region and at least one disease-susceptibility locus.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: ATRNL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat