Anti-NCAN Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-1055R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:neurocan
- Antigen symbol:NCAN
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:13004
- Antigen synonyms:C230035B04|Tgfbit|Cspg3|Cspg3-rs
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:1210-1268/1268
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
May modulate neuronal adhesion and neurite growth during development by binding to neural cell adhesion molecules (NG-CAM and N-CAM). Chondroitin sulfate proteoglycan; binds to hyaluronic acid.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NCAN
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat