Anti-HMGB1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-0664R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:High-mobility Group Box 1
- Antigen symbol:HMGB1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3146
- Antigen synonyms:high-mobility group box 1|RP11-550P23.1|high mobility group protein 1|Amphoterin|SBP-1|high-mobility group (nonhistone chromosomal) protein 1|high mobility group protein B1|HMG3|Sulfoglucuronyl carbohydrate binding protein|HMG1|HMG-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:60-110/215
- Purification:Purified by Protein A
- Pk:100 µl
DNA binding proteins that associates with chromatin and has the ability to bend DNA. Binds preferentially single-stranded DNA. Involved in V(D)J recombination by acting as a cofactor of the RAG complex. Acts by stimulating cleavage and RAG protein binding at the 23 bp spacer of conserved recombination signal sequences (RSS). Heparin-binding protein that has a role in the extension of neurite-type cytoplasmic processes in developing cells (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HMGB1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat