Eyewash bottles, Duo
Supplier: Plum Safety
Certificates
About this item
Sterile 0,9 % Sodium chloride solution is ideal for injuries with foreign bodies like dust, dirt ect. Sterile 4,9 % Phosphate buffer solution is perfect for neutralising acids and alkalis.
- Complete with an eye device for rinsing both eyes at the same time
- Also for refilling of the Duo eye wash station
Rinsing time: ~2 min. / 5 min.
Shelf life: 3 years