Order Entry
Finland
ContactUsLinkComponent
J.T.Baker® BAKERBOND Speedisk®, Extraction Stations
J.T.Baker® BAKERBOND Speedisk®, Extraction Stations
Catalog # 8095-06
Supplier:  Avantor
undefined
J.T.Baker® BAKERBOND Speedisk®, Extraction Stations
Catalog # 8095-06
Supplier:  Avantor
Specifications
  • Description:
    Extraction station, expanded, 6-port
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
The BAKERBOND Speedisk® expanded extraction station can be used in reservoir, inverted, or remote sample feed modes. It includes a six-port vacuum manifold and the accessories needed to support the extraction of analyte by BAKERBOND Speedisk® laminar extraction discs.
The manifold has a rectangular footprint and inter-port spacing to accommodate six, side-by-side, 1 L sample reservoirs. Each vacuum port has an individual open/close valve.
Delivery: Including extraction station, two remote sample adapters, two collection chambers and two vials, six reservoirs of 185 ml.