Order Entry
Finland
ContactUsLinkComponent
Anti-IKB-epsilon Rabbit Polyclonal Antibody [clone: D17 ]
Anti-IKB-epsilon Rabbit Polyclonal Antibody [clone: D17 ]
Catalog # BWRLBS1191
Anti-IKB-epsilon Rabbit Polyclonal Antibody [clone: D17 ]
Catalog # BWRLBS1191

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    IKB- epsilon
  • Clonality:
    Polyclonal
  • Clone:
    D17
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 1-50 of Human IKB-epsilon.

I?B-? (D17) pAb detects endogenous levels of I?B-? protein.

Type: Primary
Antigen: IKB-epsilon
Clonality: Polyclonal
Clone: D17
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat