Anti-SR-2C Rabbit Polyclonal Antibody [clone: P190]
BWRLBS9129
- Antibody type:Primary
- Antigen symbol:SR-2C
- Clonality:Polyclonal
- Clone:P190
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 160-210 of Human SR-2C.
SR-2C (P190) pAb detects endogenous levels of SR-2C protein.
Type: Primary
Antigen: SR-2C
Clonality: Polyclonal
Clone: P190
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat