Order Entry
Finland
ContactUsLinkComponent
Anti-MRP-L12 Rabbit Polyclonal Antibody
Anti-MRP-L12 Rabbit Polyclonal Antibody
Catalog # BWRLBS5802
Anti-MRP-L12 Rabbit Polyclonal Antibody
Catalog # BWRLBS5802

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MRP-L12
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 75-120 of Human MRP-L12.

MRP-L12 pAb detects endogenous levels of MRP-L12 protein.

Type: Primary
Antigen: MRP-L12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat