Anti-PKA Ialpha reg Rabbit Polyclonal Antibody
BWRLBS6217
- Antibody type:Primary
- Antigen symbol:PKA I alpha reg
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human PKA Ialpha reg.
PKA I? reg pAb detects endogenous levels of PKA I? reg protein.
Type: Primary
Antigen: PKA Ialpha reg
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat