Anti-p-Bad Rabbit Polyclonal Antibody [clone: S91]
Catalog # BWRLBS4474
Toimittaja: Bioworld Technology
Tekniset tiedot
- Antibody type:Primary
- Antigen symbol:p-Bad
- Clonality:Polyclonal
- Clone:S91
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Tekniset tiedot
Tietoja tästä tuotteesta
Synthetic phosphopeptide derived from human Bad around the phosphorylation site of Serine 91.
p-Bad (S91) pAb detects endogenous levels of Bad protein only when phosphorylated at Ser91.
Type: Primary
Antigen: p-Bad
Clonality: Polyclonal
Clone: S91
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat