Anti-p-FoxO1/3/4 Rabbit Polyclonal Antibody [clone: T24/32]
Catalog # BWRLBS4266
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-FoxO1/3/4
- Clonality:Polyclonal
- Clone:T24/32
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human FoxO1 around the phosphorylation site of Threonine 24.
p-FoxO1/3/4 (T24/32) pAb detects endogenous levels of FoxO1 protein only when phosphorylated at Thr24. This antibody also detects endogenous levels of FoxO3 and FoxO4 protein only when phosphorylated at Thr32.
Type: Primary
Antigen: p-FoxO1/3/4
Clonality: Polyclonal
Clone: T24/32
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat