Anti-PKA IIbeta reg Rabbit Polyclonal Antibody [clone: T109]
Catalog # BWRLBS3253
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PKA II beta reg
- Clonality:Polyclonal
- Clone:T109
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 80-130 of Human PKA IIbeta reg.
PKA II? reg (T109) pAb detects endogenous levels of PKA II? reg protein.
Type: Primary
Antigen: PKA IIbeta reg
Clonality: Polyclonal
Clone: T109
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat