Anti-SR-5A Rabbit Polyclonal Antibody [clone: L35]
BWRLBS2767
- Antibody type:Primary
- Antigen symbol:SR-5A
- Clonality:Polyclonal
- Clone:L35
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids N-terminus of Human SR-5A.
SR-5A (L35) pAb detects endogenous levels of SR-5A protein.
Type: Primary
Antigen: SR-5A
Clonality: Polyclonal
Clone: L35
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human