Order Entry
Finland
ContactUsLinkComponent
Anti-SR-2C Rabbit Polyclonal Antibody [clone: Q437]
Anti-SR-2C Rabbit Polyclonal Antibody [clone: Q437]
Catalog # BWRLBS2766
Anti-SR-2C Rabbit Polyclonal Antibody [clone: Q437]
Catalog # BWRLBS2766

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SR-2C
  • Clonality:
    Polyclonal
  • Clone:
    Q437
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 400-450 of Human SR-2C.

SR-2C (Q437) pAb detects endogenous levels of SR-2C protein.

Type: Primary
Antigen: SR-2C
Clonality: Polyclonal
Clone: Q437
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat