Anti-Rsk-4 Rabbit Polyclonal Antibody [clone: Q688]
BWRLBS2507
- Antibody type:Primary
- Antigen symbol:Rsk-4
- Clonality:Polyclonal
- Clone:Q688
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 653-700 of Human Rsk-4.
Rsk-4 (Q688) pAb detects endogenous levels of Rsk-4 protein.
Type: Primary
Antigen: Rsk-4
Clonality: Polyclonal
Clone: Q688
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human