Anti-c-Fms Rabbit Polyclonal Antibody [clone: E557]
Catalog # BWRLBS1627
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:c-Fms
- Clonality:Polyclonal
- Clone:E557
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 531-580 of Human c-Fms.
c-Fms (E557) pAb detects endogenous levels of c-Fms protein.
Type: Primary
Antigen: c-Fms
Clonality: Polyclonal
Clone: E557
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat