Anti-PKA Ibeta reg Rabbit Polyclonal Antibody [clone: F138]
Catalog # BWRLBS2269
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PKA I beta reg
- Clonality:Polyclonal
- Clone:F138
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 120-160 of Human PKA Ibeta reg.
PKA Ibeta reg (F138) pAb detects endogenous levels of PKA Ibeta reg protein.
Type: Primary
Antigen: PKA Ibeta reg
Clonality: Polyclonal
Clone: F138
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat