Anti-SR-1A Rabbit Polyclonal Antibody [clone: F321]
Catalog # BWRLBS2180
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:SR-1A
- Clonality:Polyclonal
- Clone:F321
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 291-340 of Human SR-1A.
SR-1A (F321) pAb detects endogenous levels of SR-1A protein.
Type: Primary
Antigen: SR-1A
Clonality: Polyclonal
Clone: F321
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat