Anti-OAT1 Rabbit Polyclonal Antibody [clone: L304]
BWRLBS3556
- Antibody type:Primary
- Antigen symbol:OAT1
- Clonality:Polyclonal
- Clone:L304
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 271-320 of Human OAT1.
OAT1 (L304) pAb detects endogenous levels of OAT1 protein.
Type: Primary
Antigen: OAT1
Clonality: Polyclonal
Clone: L304
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat