Anti-Neuromedin-U Rabbit Polyclonal Antibody [clone: I174]
Catalog # BWRLBS3892
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Neuromedin-U
- Clonality:Polyclonal
- Clone:I174
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 150-200 of Human Neuromedin-U.
Neuromedin-U (I174) pAb detects endogenous levels of Neuromedin-U protein.
Type: Primary
Antigen: Neuromedin-U
Clonality: Polyclonal
Clone: I174
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human