Anti-Egr-1/2 Rabbit Polyclonal Antibody [clone: R409]
Catalog # BWRLBS1944
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Egr-1/2
- Clonality:Polyclonal
- Clone:R409
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 370-420 of Human Egr-1.
Egr-2 (R409) pAb detects endogenous levels of Egr-2 protein.
Type: Primary
Antigen: Egr-1/2
Clonality: Polyclonal
Clone: R409
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat