Anti-PKA IIalpha reg Rabbit Polyclonal Antibody [clone: P68]
Catalog # BWRLBS1929
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PKA II alpha reg
- Clonality:Polyclonal
- Clone:P68
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 50-100 of Human PKA IIalpha reg.
PKA II? reg (P68) pAb detects endogenous levels of PKA II? reg protein.
Type: Primary
Antigen: PKA IIalpha reg
Clonality: Polyclonal
Clone: P68
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human