Anti-Synuclein-beta Rabbit Polyclonal Antibody [clone: S118]
BWRLBS1348
- Antibody type:Primary
- Antigen symbol:Synuclein- beta
- Clonality:Polyclonal
- Clone:S118
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 82-140 of Human Synuclein-beta.
Synuclein ? (S118) pAb detects endogenous levels of Synuclein ? protein.
Type: Primary
Antigen: Synuclein-beta
Clonality: Polyclonal
Clone: S118
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat