Food DNA kit, E.Z.N.A.®
The E.Z.N.A.® Food DNA Kit allows rapid and reliable isolation of high quality DNA from complex matrixes such as processed food, chocolate, cereals and meat.
- Quality: Purified DNA suitable for a variety of sensitive downstream applications
- Safe: No organic extractions
- Versatile: Isolate DNA from a range of food products including milk, cereal and chocolate
The system combines HiBind® DNA mini columns with RBB Buffer to eliminate PCR inhibiting compounds within the samples and elute highly concentrated DNA. The purified DNA can be used in PCR-based testing for genetically modified organism (GMO) DNA.
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Food samples
- Target molecule:DNA
- Sample size:200 mg food or 0,5 ml milk
- Description:E.Z.N.A.® Food DNA Kit