Cell culture dishes, cell-repellent surface, CELLSTAR®
Supplier: Greiner Bio-One
Certificates
About this item
These sterile, PS cell culture dishes have a cell-repellent surface to effectively inhibit cell attachment. Achieved through a chemical polymer modification, the surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
- Effectively inhibits cell adhesion
- Sterile
- Lid with condensation rings
- Other formats available (flasks, microplates, multiwell plates)
Characterised by low cell attachment, the cell-repellent surface is ideal for applications such as spheroid cultures and stem cell aggregate formation, suspension culture of semi-adherent and adherent cell (eg. for suspension culture of macrophages), gel-based 3D cultures, or magnetic 3D cell culturing (Nano3D technology).
Specifications
- Catalog No:
- GREI664970_5
- GREI627979_40
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)