

Ethanol absolute, EMPROVE® EXPERT BP, Ph. Eur., USP suitable for use as excipient
:64-17-5
:
:C₂H₅OH
:MFCD00003568
:46.07 g/mol
:1170
:3,II
:GHS02
:GHS07
:Freezer
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Purity (GC):99,8%
- Assay (m/m):99,2%
- Assay (V/V):99,5%
- Indentity (IR):Conforms
- Appearance:Conforms
- Colour:≤10 Hazan
- Clarity of solution:Conforms
- Colour of solution:Conforms
- Acidity or alkalinity:≤30 ppm
- Acidity (as CH₃COOH):≤30 ppm
- Refractive Index (n 25/D):1,3589 - 1,3599
- Density (d 20 °C/20 °C):0,790 - 0,793
- Density (d 15,56 °C/15,56 °C):≤0,7962
- Absorbance (at 240 nm):≤0,40
- Absorbance (between 250 and 260 nm):≤0,30
- Absorbance (between 270 and 340 nm):≤0,10
- Volatile impurities (GC; acetal and acetaldehyde):≤10 ppm
- Volatile impurities (GC; benzene):≤2 ppm
- Volatile impurities (GC; methanol):≤100 ppm
- Volatile impurities (GC; total-other impurities):≤300 ppm
- Volatile impurities (GC; disregard limit):≤9 ppm
- Cyclohexane (GC):≤50 ppm
- 2-Propanol (GC):≤30 ppm
- Evaporation residue:≤0,0001%
- Water:≤0,500%
- Bacterial endotoxins:≤2,5 IU/ml
- Total areobic microbial unit (TAMC):≤100 CFU/g
- Total combained yeasts/moulds count (TYMC):≤10 CFU/g
- Bile-tolerant gram negative bacteria (absent in 1 g):Passes Test
- Candida albicans (absent in 1 g):Passes Test
- Escherichia coli (absent in 1 g):Passes Test
- Pseudomonas aeruginosa (absent in 1 g):Passes Test
- Salmonella (absent in 10 g):Passes Test
- Staphylococcus aureus (absent in 1 g):Passes Test
Frequently Bought Together