Order Entry
Finland
ContactUsLinkComponent
Dextran sulphate sodium salt, M.W. ∼ 400,000-600,000, white powder
Dextran sulphate sodium salt, M.W. ∼ 400,000-600,000, white powder
Supplier:  MP Biomedicals
Certificates
Dextran sulphate sodium salt, M.W. ∼ 400,000-600,000, white powder
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • FTIR:
    Conforms to Standard
  • pH (1% aq soln):
    5 - 7,2
  • Loss on Drying:
    <10%
  • Optical Rotation (c=1, water):
    +75° to +105°

Specification Test Results