Silicon dioxide 200-500 µm
Catalog # ICNA0202809
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:60.08 g/mol
Storage Temperature:Ambient
CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232