Drums, wide mouth, UN approved
Supplier: LAMAPLAST
Certificates
About this item
Drums, made of HDPE, blue, UN approved, with lid.
- Stackable
- Tamper evident
- Self-sealing lid with zinc plate steel closure
- For food use
For food, cosmetic and chemical fields. Manufactured with the inner neutral layer that avoids the contact between the content and the coloring pigment.
Dimensions allow overlap on pallet 800×1200 and 1000×1200.