Detergents, neodisher® MediClean
Supplier: Dr. Weigert
Certificates
About this item
neodisher® MediClean is a detergent with slightly alkaline active cleaning component in a low form formulation.
- Liquid concentrate for the automated cleaning of thermostable and thermolabile instruments
- Components: <5% anionic surfactants also enzymes, preservatives (phenoxyethanol, methyl-, ethyl-, butyl-, propyl-, isopropylparaben)
- Dosage: 0,2 to 1,0%