Chemical resistant boot covers, MICROGARD® 2500, model 406
ANSEWH25B-00406-00
: Ansell
- Shoe size:42 - 46
- Garment application:Chemical protection,Laboratory
- Colour:White
- Pk:200 PAIR
These over boots are made of microporous PP film with a non woven inner layer. The fabric provides an excellent barrier to chemical spray and infective agents.
- Elastic to top of boot
- Tie fastening
- Bound seams
- Fit shoe size 42 - 46