Corning® ITS Universal Culture Supplement Premix, Corning
Supplier: Corning
Certificates
About this item
ITS contains insulin, human transferrin, and selenous acid, the three most universally essential components of defined culture media.
They stimulate cell proliferation of a variety of cells under serum-reduced conditions.
Tested for ability to promote proliferation of HeLa cells in serum-free medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi and mycoplasma.
Specifications
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 14001 - Bedford
- Formulation:As an aqueous solution containing human recombinant insulin, human transferrin (12,5 mg each), selenous acid
(12,5 µg), BSA (2.5 g) and linoleic acid (10,7 mg) - Quality control:Tested for ability to promote proliferation of HeLa cells in serum-free medium
Filtered (0.2 µm membrane) and tested and found negative for bacteria, fungi and mycoplasma - Recommended dilution:1:1000 (dilute stock solution in serum-free medium)
- Storage and stability:Stable for at least three months at 2 to 8 °C. Do not freeze.