Genomic DNA isolation from blood, E.Z.N.A.® SQ blood DNA kit
Catalog # D5032-03
Supplier: OMEGA BIO-TEK
CAS Number:
Specifications
- Pk:300 mL
- Method / Format:Precipitation
- Prep size:Scalable
- Refine target molecule:Total DNA
- Sample type:Blood
- Target molecule:DNA
- Description:E.Z.N.A.® SQ Blood DNA Kit
- Sample size:Up to 20 ml
- Description:E.Z.N.A.® SQ Blood DNA Kit
Specifications
About this item
The E.Z.N.A.® SQ blood DNA kit provides a reliable method for the isolation of high molecular weight genomic DNA from fresh, frozen, or anti-coagulated whole blood. The method can also be used for preparation of genomic DNA from buffy coat, bone marrow or cultured cells. There is no need for phenol/chloroform extractions, and time consuming steps such as CsCl gradient ultracentrifugation are eliminated. DNA purified using the E.Z.N.A.® SQ blood DNA method is ready for downstream applications such as PCR, southern blotting and restriction enzyme digestion.
- Isolation of high quality, high molecular weight genomic DNA
- Versatility in amount of starting material, with DNA purification from 0,1 to 10 ml whole blood
- Process single or multiple samples in less than 90 minutes