Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Glycyrrhetic acid
Glycyrrhetic acid ≥98%
Glycyrrhetic acid ≥98%
Glycyrrhetic acid ≥98%
Supplier:
Cayman Chemical
Certificates
Expand image
Glycyrrhetic acid ≥98%
Supplier:
Cayman Chemical
Certificates
About this item
CAS Number:
471-53-4
:
Formula:
C₃₀H₄₆O₄
MDL Number:
MFCD00003706
:
470.69 g/mol
Storage Temperature:
Freezer
Related Information
Documentation
Certificates
Search by Lot Number
Documentation