Toray carbon paper, TGP-H-60
Toray carbon paper, TGP-H-60 can be used as a gas diffusion layer in fuel cell applications. It is also suitable for use as a catalyst backing layer.
- Insoluble in water
- Air and light sensitive
- Container must be closed tightly and stored in a cool, dry and well-ventilated place
It is incompatible with strong oxidizing agents, chlorinated paraffins, lead oxide, manganese oxide, iron oxide, liquid oxygen, oils, chlorinetrifluoride, fluoride and potassium superoxide.
: Available in PTFE treated version.